trans-4-Hexen-1-ol 98% (E> 85%)
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-Hex-4-en-1-ol, trans-Hex-4-en-1-ol
A1360391-25G
A1360391-5G
76659-012EA
68.96
USD
76659-012
76659-014
trans-4-Hexen-1-ol 98% (E> 85%)
trans-4-Hexen-1-ol
Precaución: for Research Use Only
Formula:
C₆H₁₂O MW: 100.16 g/mol |
MDL Number:
MFCD00009713 CAS Number: 928-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...