(R)-2-((tert-Butoxycarbonyl)amino)non-8-enoic acid 95%

Supplier: Ambeed
Warning

A1360381-1G A1360381-5G
76659-008EA 269.47 USD
76659-008 76659-010
(R)-2-((tert-Butoxycarbonyl)amino)non-8-enoic acid 95%
(R)-2-((tert-Butoxycarbonyl)amino)non-8-enoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₄
MW: 271.35 g/mol
MDL Number: MFCD22422689
CAS Number: 881683-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR