2-(4'-(Diphenylamino)-[1,1'-biphenyl]-4-yl)acetonitrile 98%

Supplier: Ambeed
Warning

A1359677-100MG A1359677-1G A1359677-250MG
76658-984EA 16.98 USD
76658-984 76658-986 76658-988
2-(4'-(Diphenylamino)-[1,1'-biphenyl]-4-yl)acetonitrile 98%
2-(4'-(Diphenylamino)-[1,1'-biphenyl]-4-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₂
MW: 360.4504 g/mol
CAS Number: 1000504-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR