4-Methyl-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N-(p-tolyl)aniline 98%

Supplier: Ambeed
Warning

A1359674-1G A1359674-250MG
76658-980EA 60.48 USD
76658-980 76658-982
4-Methyl-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N-(p-tolyl)aniline 98%
4-Methyl-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N-(p-tolyl)aniline

Precaución: for Research Use Only

Formula: C₂₆H₃₀BNO₂
MW: 399.3329 g/mol
CAS Number: 528610-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR