N-(4-(4-Methylpiperidin-1-yl)-3-(trifluoromethyl)phenyl)-4-(morpholinomethyl)benzamide 97%

Supplier: Ambeed
Warning

A1359565-10MG A1359565-1MG A1359565-25MG A1359565-50MG A1359565-5MG A1359565-100MG
76658-964EA 42 USD
76658-964 76658-966 76658-968 76658-970 76658-972 77867-522
N-(4-(4-Methylpiperidin-1-yl)-3-(trifluoromethyl)phenyl)-4-(morpholinomethyl)benzamide 97%
N-(4-(4-Methylpiperidin-1-yl)-3-(trifluoromethyl)phenyl)-4-(morpholinomethyl)benzamide

Precaución: for Research Use Only

Formula: C₂₅H₃₀F₃N₃O₂
CAS Number: 2479465-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR