2-Nitro-[1,1'-biphenyl]-4,4'-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A1359549-100MG A1359549-250MG
76658-960EA 40 USD
76658-960 76658-962
2-Nitro-[1,1'-biphenyl]-4,4'-dicarboxylic acid 97%
2-Nitro-[1,1'-biphenyl]-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₉NO₆
CAS Number: 1314484-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR