2-(9-(Pyridin-2-yl)-6-oxaspiro[4.5]decan-9-yl)acetonitrile 97%

Supplier: Ambeed
Warning

A1359547-1G A1359547-5G
76658-958EA 129 USD
76658-958 77867-521
2-(9-(Pyridin-2-yl)-6-oxaspiro[4.5]decan-9-yl)acetonitrile 97%
2-(9-(Pyridin-2-yl)-6-oxaspiro[4.5]decan-9-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O
CAS Number: 1401031-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR