(2-Chloro-10H-phenothiazin-10-yl)(4-methoxyphenyl)methanone 98%

Supplier: Ambeed
Warning

Synonyms: Tubulin inhibitor 6

A1359471-10MG A1359471-1MG A1359471-5MG
76658-946EA 36 USD
76658-946 76658-948 76658-950
(2-Chloro-10H-phenothiazin-10-yl)(4-methoxyphenyl)methanone 98%
(2-Chloro-10H-phenothiazin-10-yl)(4-methoxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₂₀H₁₄ClNO₂S
MDL Number: MFCD01245092
CAS Number: 105925-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR