(R)-2-(Adamantan-1-yl)-2-aminoacetic acid hydrochloride 97%

Supplier: Ambeed
Warning

A1359411-250MG A1359411-1G
76658-912EA 96 USD
76658-912 77621-714
(R)-2-(Adamantan-1-yl)-2-aminoacetic acid hydrochloride 97%
(R)-2-(Adamantan-1-yl)-2-aminoacetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₀ClNO₂
CAS Number: 101976-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR