rel-(1R,3S)-Methyl 3-hydroxycyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

A1359399-100MG A1359399-250MG
76658-908EA 53 USD
76658-908 76658-910
rel-(1R,3S)-Methyl 3-hydroxycyclohexanecarboxylate 97%
rel-(1R,3S)-Methyl 3-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD27926393
CAS Number: 6183-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR