Methyl 3-(2,4-dichlorophenyl)-2-hydroxypropanoate 97%

Supplier: Ambeed
Warning

A1359090-1G A1359090-250MG
76658-896EA 280 USD
76658-896 76658-898
Methyl 3-(2,4-dichlorophenyl)-2-hydroxypropanoate 97%
Methyl 3-(2,4-dichlorophenyl)-2-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₃
MDL Number: MFCD21331191
CAS Number: 875018-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR