Dimethyl 4,5-dimethylphthalate 98%

Supplier: Ambeed
Warning

A1355919-1G A1355919-250MG
76658-872EA 124.13 USD
76658-872 76658-874
Dimethyl 4,5-dimethylphthalate 98%
Dimethyl 4,5-dimethylphthalate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.2372 g/mol
CAS Number: 17649-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR