4-Chloro-2-(2,6-dioxopiperidin-3-yl)-1H-pyrrolo[3,4-c]pyridine-1,3(2H)-dione 98%
Supplier: Ambeed
|
Warning
|
A1355843-100MG
A1355843-1G
A1355843-250MG
76658-866EA
23
USD
76658-866
76658-868
76658-870
4-Chloro-2-(2,6-dioxopiperidin-3-yl)-1H-pyrrolo[3,4-c]pyridine-1,3(2H)-dione 98%
4-Chloro-2-(2,6-dioxopiperidin-3-yl)-1H-pyrrolo[3,4-c]pyridine-1,3(2H)-dione
Precaución: for Research Use Only
Formula:
C₁₂H₈ClN₃O₄ |
CAS Number:
2154357-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...