2-((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)-2-((S)-4-phenyloxazolidin-2-ylidene)acetonitrile 98%

Supplier: Ambeed
Warning

A1355479-100MG A1355479-1G A1355479-250MG A1355479-5G
76658-860EA 32 USD
76658-860 76658-862 76658-864 77621-696
2-((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)-2-((S)-4-phenyloxazolidin-2-ylidene)acetonitrile 98%
2-((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)-2-((S)-4-phenyloxazolidin-2-ylidene)acetonitrile

Precaución: for Research Use Only

Formula: C₂₀H₁₇N₃O₂
MDL Number: MFCD00192394
CAS Number: 150639-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR