(4-(Pyridin-2-yl)phenyl)methanamine dihydrochloride 98%

Supplier: Ambeed
Warning

A1355020-1G A1355020-250MG A1355020-5G
76658-838EA 47 USD
76658-838 76658-840 76658-842
(4-(Pyridin-2-yl)phenyl)methanamine dihydrochloride 98%
(4-(Pyridin-2-yl)phenyl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂N₂
MW: 257.1590 g/mol
MDL Number: MFCD31560240
CAS Number: 2470440-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR