4-Hydrazinyl-1-methyl-1H-pyrazole hydrochloride 98%

Supplier: Ambeed
Warning

A1354975-100MG A1354975-250MG
76658-834EA 132.62 USD
76658-834 76658-836
4-Hydrazinyl-1-methyl-1H-pyrazole hydrochloride 98%
4-Hydrazinyl-1-methyl-1H-pyrazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉ClN₄
MW: 148.5941 g/mol
CAS Number: 1959556-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR