(2R,3S,5R)-5-(4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol 97%

Supplier: Ambeed
Warning

A1354820-100MG A1354820-1G A1354820-250MG A1354820-50MG
76658-826EA 137.92 USD
76658-826 76658-828 76658-830 76658-832
(2R,3S,5R)-5-(4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol 97%
(2R,3S,5R)-5-(4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-1-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂IN₅O₃
MW: 377.1385 g/mol
CAS Number: 187479-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR