6-Methoxy-2-(4-methoxyphenyl)-1H-indole-3-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A1354770-100MG
A1354770-5G
76658-822EA
94.43
USD
76658-822
76658-824
6-Methoxy-2-(4-methoxyphenyl)-1H-indole-3-carbaldehyde 97%
6-Methoxy-2-(4-methoxyphenyl)-1H-indole-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₇H₁₅NO₃ MW: 281.3059 g/mol |
CAS Number:
158611-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...