Riboflavine-5'-phosphate sodium salt dihydrate 73-79%

Supplier: Ambeed
Warning

A1353492-100G A1353492-1G A1353492-25G A1353492-5G
76658-800EA 124 USD
76658-800 76658-802 76658-804 76658-806
Riboflavine-5'-phosphate sodium salt dihydrate 73-79%
Riboflavine-5'-phosphate sodium salt dihydrate

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₄NaO₉P·2H₂O
MDL Number: MFCD00150993
CAS Number: 6184-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR