1-(2-Aminoethyl)cyclohexanol hydrochloride 97%

Supplier: Ambeed
Warning

A1353309-1G A1353309-250MG
76658-796EA 326.76 USD
76658-796 76658-798
1-(2-Aminoethyl)cyclohexanol hydrochloride 97%
1-(2-Aminoethyl)cyclohexanol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈ClNO
MW: 179.6876 g/mol
MDL Number: MFCD13176553
CAS Number: 2155856-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR