trans-m-Coumaric acid 97%

Supplier: Ambeed
Warning

A1353299-5G A1353299-25G A1353299-100G
76658-794EA 11 USD
76658-794 77596-844 77867-469
trans-m-Coumaric acid 97%
trans-m-Coumaric acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD00004390
CAS Number: 14755-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR