2,6-Bis((S)-4-cyclohexyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A1353132-1G A1353132-250MG A1353132-100MG
76658-782EA 393 USD
76658-782 76658-784 77621-684
2,6-Bis((S)-4-cyclohexyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((S)-4-cyclohexyl-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₂₃H₃₁N₃O₂
CAS Number: 1562372-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR