(R)-4-(tert-Butyl)-2-(pyrimidin-2-yl)-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1353096-100MG A1353096-1G A1353096-250MG
76658-776EA 65.78 USD
76658-776 76658-778 76658-780
(R)-4-(tert-Butyl)-2-(pyrimidin-2-yl)-4,5-dihydrooxazole 98%
(R)-4-(tert-Butyl)-2-(pyrimidin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O
MW: 205.2563 g/mol
CAS Number: 2757082-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR