(R)-4-Benzyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

A1353088-100MG A1353088-1G A1353088-250MG
76658-756EA 28 USD
76658-756 76658-758 76658-760
(R)-4-Benzyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%
(R)-4-Benzyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₂O
CAS Number: 2757082-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR