(R)-4-Isopropyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

A1353086-100MG A1353086-1G A1353086-250MG
76658-750EA 42 USD
76658-750 76658-752 76658-754
(R)-4-Isopropyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%
(R)-4-Isopropyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O
CAS Number: 2757082-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR