(R)-4-(tert-Butyl)-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

A1353085-100MG A1353085-1G A1353085-250MG
76658-744EA 63 USD
76658-744 76658-746 76658-748
(R)-4-(tert-Butyl)-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%
(R)-4-(tert-Butyl)-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂O
CAS Number: 2757082-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR