(R)-2-(1,8-Naphthyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%
Supplier: Ambeed
|
Warning
|
A1353074-100MG
A1353074-1G
A1353074-250MG
76658-728EA
159.14
USD
76658-728
76658-730
76658-732
(R)-2-(1,8-Naphthyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%
(R)-2-(1,8-Naphthyridin-2-yl)-4-phenyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₇H₁₃N₃O MW: 275.3046 g/mol |
CAS Number:
2757082-69-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...