(3aR,8aS)-2-(1,8-Naphthyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole 98%

Supplier: Ambeed
Warning

A1353067-100MG A1353067-1G A1353067-250MG
76658-694EA 148.53 USD
76658-694 76658-696 76658-698
(3aR,8aS)-2-(1,8-Naphthyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole 98%
(3aR,8aS)-2-(1,8-Naphthyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole

Precaución: for Research Use Only

Formula: C₁₈H₁₃N₃O
MW: 287.3153 g/mol
CAS Number: 2757082-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR