6,6'-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A1353062-100MG A1353062-1G A1353062-250MG
76658-682EA 132.62 USD
76658-682 76658-684 76658-686
6,6'-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥98%, ee 99%
6,6'-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₃₀H₂₆N₄O₂
MW: 474.5531 g/mol
CAS Number: 2757082-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR