(4R,5R)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

A1353055-100MG A1353055-1G A1353055-250MG
76658-652EA 148.53 USD
76658-652 76658-654 76658-656
(4R,5R)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole ≥98%, ee 99%
(4R,5R)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₁₉N₃O
MW: 377.4379 g/mol
CAS Number: 2757082-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR