(4R,5S)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole 98%
Supplier: Ambeed
|
Warning
|
A1353053-100MG
A1353053-1G
A1353053-250MG
A1353053-50MG
76658-638EA
146.41
USD
76658-638
76658-640
76658-642
76658-644
(4R,5S)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole 98%
(4R,5S)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₂₅H₁₉N₃O MW: 377.4379 g/mol |
CAS Number:
2828432-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...