(4R,4'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 98%

Supplier: Ambeed
Warning

A1353051-100MG A1353051-1G A1353051-250MG
76658-632EA 78 USD
76658-632 76658-634 76658-636
(4R,4'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 98%
(4R,4'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₉H₃₈N₂O₂
MW: 446.6242 g/mol
CAS Number: 2828432-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR