(4S,4'S)-4,4'-Di-sec-butyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 98%

Supplier: Ambeed
Warning

A1353047-1G A1353047-250MG A1353047-100MG
76658-626EA 397 USD
76658-626 76658-628 77621-680
(4S,4'S)-4,4'-Di-sec-butyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 98%
(4S,4'S)-4,4'-Di-sec-butyl-4,4',5,5'-tetrahydro-2,2'-bioxazole

Precaución: for Research Use Only

Formula: C₁₄H₂₄N₂O₂
CAS Number: 2374958-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR