(4S,4'S)-4,4'-Diisobutyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 98%

Supplier: Ambeed
Warning

A1353040-100MG A1353040-1G
76658-622EA 140.04 USD
76658-622 76658-624
(4S,4'S)-4,4'-Diisobutyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 98%
(4S,4'S)-4,4'-Diisobutyl-4,4',5,5'-tetrahydro-2,2'-bioxazole

Precaución: for Research Use Only

Formula: C₁₄H₂₄N₂O₂
MW: 252.3526 g/mol
CAS Number: 138429-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR