4-Chloro-5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde 97%

Supplier: Ambeed
Danger

A1352957-1G A1352957-250MG
76658-618EA 164.44 USD
76658-618 76658-620
4-Chloro-5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde 97%
4-Chloro-5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₇ClFNO
MW: 223.6308 g/mol
MDL Number: MFCD27933304
CAS Number: 928324-56-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR