2-(((9H-Purin-6-yl)amino)methyl)-6-methylphenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
PI-55
A1352831-100MG
A1352831-1G
A1352831-250MG
A1352831-50MG
76658-600EA
326.76
USD
76658-600
76658-602
76658-604
77621-672
2-(((9H-Purin-6-yl)amino)methyl)-6-methylphenol 98%
2-(((9H-Purin-6-yl)amino)methyl)-6-methylphenol
Precaución: for Research Use Only
Formula:
C₁₃H₁₃N₅O MW: 255.28 g/mol |
MDL Number:
MFCD34165679 CAS Number: 1122579-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...