4-Hydroxy-3-methylpyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A1352821-100MG A1352821-1G A1352821-250MG
76658-594EA 95 USD
76658-594 76658-596 76658-598
4-Hydroxy-3-methylpyridin-2(1H)-one 95%
4-Hydroxy-3-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MDL Number: MFCD13175383
CAS Number: 4664-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR