4,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A1352789-100MG A1352789-1G A1352789-250MG
76658-588EA 150.65 USD
76658-588 76658-590 76658-592
4,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 97%
4,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₃₂H₂₆N₂O₃
MW: 486.5605 g/mol
CAS Number: 1080596-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR