2-Pyridinethiol 97%

Supplier: Ambeed
Warning

A1352707-25G A1352707-500G A1352707-100G
76658-574EA 9 USD
76658-574 76658-576 77592-558
2-Pyridinethiol 97%
2-Pyridinethiol

Precaución: for Research Use Only

Formula: C₅H₅NS
MDL Number: MFCD00006285
CAS Number: 2637-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR