(2-Chlorophenyl)diphenylphosphine 97%

Supplier: Ambeed
Warning

A1352609-100MG A1352609-1G A1352609-250MG
76658-558EA 43.5 USD
76658-558 76658-560 76658-562
(2-Chlorophenyl)diphenylphosphine 97%
(2-Chlorophenyl)diphenylphosphine

Precaución: for Research Use Only

Formula: C₁₈H₁₄ClP
MW: 296.7305 g/mol
MDL Number: MFCD03284243
CAS Number: 35035-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR