N1-Dodecylpropane-1,3-diamine 98%

Supplier: Ambeed
Danger

A1352346-1G A1352346-250MG
76658-552EA 107.16 USD
76658-552 77549-628
N1-Dodecylpropane-1,3-diamine 98%
N1-Dodecylpropane-1,3-diamine

Precaución: for Research Use Only

Formula: C₁₅H₃₄N₂
MW: 242.44 g/mol
MDL Number: MFCD00014828
CAS Number: 5538-95-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR