5-Bromo-7-methylbenzo[b]thiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A1352326-1G A1352326-250MG A1352326-5G
76658-542EA 77 USD
76658-542 76658-544 76658-546
5-Bromo-7-methylbenzo[b]thiophene-2-carboxylic acid 97%
5-Bromo-7-methylbenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇BrO₂S
MDL Number: MFCD29784001
CAS Number: 1935615-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR