(R)-2-(2-(Diphenylphosphanyl)phenyl)-4-isobutyl-4,5-dihydrooxazole 98+%

Supplier: Ambeed
Warning

A1352236-100MG A1352236-1G A1352236-250MG
76658-530EA 98 USD
76658-530 76658-532 76658-534
(R)-2-(2-(Diphenylphosphanyl)phenyl)-4-isobutyl-4,5-dihydrooxazole 98+%
(R)-2-(2-(Diphenylphosphanyl)phenyl)-4-isobutyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₂₆NOP
MW: 387.4538 g/mol
CAS Number: 2757082-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR