(3aS,8aR)-2-(6-Phenylpyridin-2-yl)-8,8a-dihydro-3aH-indeno[1,2-d]oxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1352228-100MG A1352228-1G A1352228-250MG A1352228-50MG
76658-518EA 94 USD
76658-518 76658-520 76658-522 76658-524
(3aS,8aR)-2-(6-Phenylpyridin-2-yl)-8,8a-dihydro-3aH-indeno[1,2-d]oxazole ≥97%, ee 99%
(3aS,8aR)-2-(6-Phenylpyridin-2-yl)-8,8a-dihydro-3aH-indeno[1,2-d]oxazole

Precaución: for Research Use Only

Formula: C₂₁H₁₆N₂O
CAS Number: 2634687-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR