Tris(2,2'-bipyridine)nickel(II) bromide 95%

Supplier: Ambeed
Danger

A1352224-1G A1352224-25G A1352224-5G A1352224-100G
76658-508EA 27 USD
76658-508 76658-510 76658-512 77867-453
Tris(2,2'-bipyridine)nickel(II) bromide 95%
Tris(2,2'-bipyridine)nickel(II) bromide

Precaución: for Research Use Only

Formula: C₃₀H₂₄Br₂N₆Ni
CAS Number: 15376-08-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR