(R)-2-(2-(Diphenylphosphanyl)phenyl)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1352080-100MG A1352080-1G A1352080-250MG
76658-484EA 149.59 USD
76658-484 76658-486 76658-488
(R)-2-(2-(Diphenylphosphanyl)phenyl)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazole 98%
(R)-2-(2-(Diphenylphosphanyl)phenyl)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₃₆H₃₂NOP
MW: 525.6192 g/mol
CAS Number: 1904575-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR