(4S,5R)-2-(2-(Diphenylphosphanyl)phenyl)-4,5-diphenyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1351982-100MG A1351982-1G A1351982-250MG
76658-472EA 140.04 USD
76658-472 76658-474 76658-476
(4S,5R)-2-(2-(Diphenylphosphanyl)phenyl)-4,5-diphenyl-4,5-dihydrooxazole 97%
(4S,5R)-2-(2-(Diphenylphosphanyl)phenyl)-4,5-diphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₃₃H₂₆NOP
MW: 483.5394 g/mol
CAS Number: 2468233-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR