(S)-4-Cyclohexyl-2-(2-(diphenylphosphanyl)phenyl)-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1351980-250MG A1351980-1G
76658-470EA 275 USD
76658-470 77867-448
(S)-4-Cyclohexyl-2-(2-(diphenylphosphanyl)phenyl)-4,5-dihydrooxazole 98%
(S)-4-Cyclohexyl-2-(2-(diphenylphosphanyl)phenyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₇H₂₈NOP
CAS Number: 2634687-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR