(S)-2-([2,2'-Bipyridin]-6-yl)-4-(tert-butyl)-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1351979-100MG A1351979-1G A1351979-250MG
76658-464EA 130.5 USD
76658-464 76658-466 76658-468
(S)-2-([2,2'-Bipyridin]-6-yl)-4-(tert-butyl)-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-([2,2'-Bipyridin]-6-yl)-4-(tert-butyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O
MW: 281.3523 g/mol
CAS Number: 2095304-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR