(4S,5R)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%
Supplier: Ambeed
|
Warning
|
A1351977-100MG
A1351977-1G
A1351977-250MG
76658-452EA
84
USD
76658-452
76658-454
76658-456
(4S,5R)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole 97%
(4S,5R)-2-([2,2'-Bipyridin]-6-yl)-4,5-diphenyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₂₅H₁₉N₃O |
CAS Number:
2828432-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...